Learn More
Selleck Chemical LLC Y15-25mg
SDP
Catalog No. 502389699
Encompass_Preferred
Y15 (1,2,4,5-Benzenetetraamine tetrahydrochloride, FAK inhibitor 14) is a small-molecule FAK scaffolding inhibitor that directly inhibits FAK autophosphorylation in a dose- and time-dependent manner.
Catalog No. 50-238-9699 Supplier Selleck Chemical LLC Supplier No. S532125MG
Y15 (1,2,4,5-Benzenetetraamine tetrahydrochloride, FAK inhibitor 14) is a small-molecule FAK scaffolding inhibitor that directly inhibits FAK autophosphorylation in a dose- and time-dependent manner.