Learn More
Medchemexpress LLC Y15 (FAK inhibitor 14) | 4506-66-5 | 98.0% | 284.01 g/mol | C6H14Cl4N4 | 100MG
SDP
Catalog No. 5000236263
Encompass_Preferred
Y15 (FAK Inhibitor 14) is a small-molecule research compound that inhibits focal adhesion kinase (FAK) autophosphorylation, reduces cancer cell viability in vitro, and has been used to block tumor growth in preclinical studies. It is supplied as a research-grade reagent for cellular and biochemical assays.
Catalog No. 50-002-36263 Supplier Medchemexpress LLC Supplier No. HY12444100MG
Y15 (FAK Inhibitor 14) is a small-molecule research compound that inhibits focal adhesion kinase (FAK) autophosphorylation, reduces cancer cell viability in vitro, and has been used to block tumor growth in preclinical studies. It is supplied as a research-grade reagent for cellular and biochemical assays.