Learn More
Reprocell Y27632 2MG IN SOLUTION
Catalog No. NC0454602
Encompass
Y27632 is a cell-permeable small molecule Rho-associated kinase (ROCK) inhibitor1. Y27632 has been found to prevent apoptosis as well as enhance the survival and cloning efficiency of dissociated human embryonic stem (ES) cells without affecting their self-renewal properties or pluripotency2. This molecule has also been shown to enhance survival during the transplantation of ES cell-derived neural precursors3. Y27632 in combination with Pifithrin-a� significantly improves cell recovery after cryopreservation4.
Catalog No. NC0454602 Supplier Reprocell Supplier No. 04001202
Y27632 is a cell-permeable small molecule Rho-associated kinase (ROCK) inhibitor1. Y27632 has been found to prevent apoptosis as well as enhance the survival and cloning efficiency of dissociated human embryonic stem (ES) cells without affecting their self-renewal properties or pluripotency2. This molecule has also been shown to enhance survival during the transplantation of ES cell-derived neural precursors3. Y27632 in combination with Pifithrin-a� significantly improves cell recovery after cryopreservation4.