Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Y5 ≥99%

Catalog No. 912751250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-275-1250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 91-275-1250MG Supplier Sigma Aldrich Supplier No. 912751250MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Non-fullerene acceptors (NFAs) are currently a major focus of research in the development of bulk-heterojunction organic solar cells (OSCs). In contrast to the widely used fullerene acceptors (FAs), the optical properties and electronic energy levels of NFAs can be designed and readily tuned. NFA-based OSCs can also achieve greater thermal stability and photochemical stability, as well as longer device lifetimes, than their FA-based counterparts. Recent developments have led to a rapid increase in power conversion efficiencies for NFA OSCs, with values now exceeding 15% in a single junction cell, and >17% for a tandem cell, demonstrating the viability of using NFAs to replace FAs in next-generation high-performance OSCs.Y5 is a high-performance non-fullerene acceptor (NFA) that uses an electron-deficient molecular core to obtain a low bandgap NFA and high absorption coefficient. OPV devices made from Y5 can reach high performances in single-junction devices. Y5 is versatile and can be employed in both conventional and inverted OPV device architectures and has reported PCE from 10.5 to 14.1% when paired with five different donor polymers.

Specifications

CAS 2304444-48-0
Molecular Formula C82H90N8O2S5
Linear Formula C82H90N8O2S5
Quantity 250 mg
Synonym (2,2 -((2 Z,2 Z)-((12,13-bis(2-ethylhexyl)-3,9-diundecyl-12,13-dihydro[1,2,5 ]thiadiazolo[3,4 e]thieno[2 ,3 :4 ,5 ] thieno[2 ,3 :4,5 ]pyrrolo[3,2-g] thieno[2 ,3 :4,5 ]thieno[3,2-b]indole-2,10-diyl)bis(methanylylidene))bis(3-oxo-2,3-dihydro1H-indene-2,1-diylidene))dimalononitrile)
Molecular Weight (g/mol) 1379.97
Percent Purity ≥99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.