Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Y5, Thermo Scientific Chemicals

Catalog No. H66460ME
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H66460-ME 500 mg
H66460-MC 100 mg
H66460-MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H66460-ME Supplier Thermo Scientific Chemicals Supplier No. H66460ME
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2304444-48-0
Molecular Formula C{8}{2}H{9}{0}N{8}O{2}S{5}
Synonym (2,2’-((2 Z,2’Z)-((12,13-bis(2-ethylhexyl)-3,9-diundecyl-12,13-dihydro[1,2,5 ]thiadiazolo[3,4 e]thieno[2’’,3’’:4’,5’] thieno[2’,3’:4,5 ]pyrrolo[3,2-g] thieno[2’,3’:4,5 ]thieno[3,2-b]indole-2,10-diyl)bis(methanylylidene))bis(3-oxo-2,3-dihydro1H-indene-2,1-diylidene))dimalononitrile)

Specifications

Quantity 500 mg
Formula Weight 1379.97
Physical Form Solid
Chemical Name or Material Y5
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.