Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Y7 ≥99%

Catalog No. 913251100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-325-1100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 91-325-1100MG Supplier Sigma Aldrich Supplier No. 913251100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Synonym: 2,2-((2Z,2Z)-((12,13-bis(2-ethylhexyl)-3,9-diundecyl-12,13-dihydro-[1,2,5]thiadiazolo[3,4-e]thieno[2,3:4,5]-thieno[2,3:4,5]pyrrolo[3,2-g]thieno[2,3:4,5]thieno[3,2-b]indole-2,10-diyl)bis(methanylylidene))bis(5,6-dichloro-3-oxo-2,3-dihydro-1H-indene-2,1-diylidene))dimalononitrileBTP-4Cl is a high performance, low bandgap, non-fullerene accpetor (NFA). It is the chlorinated derivative of the fused benzothiadiazole-based NFA, Y6. It offer high power conversion efficiency (PCE) when pairing with p-type polymer PM6. A 9 mm2 single junction device of this blend provided a PCE of 16.5% at thickness of ∼ 100nm, and provided a PCE of >13% at a thickness of ∼300 nm (for the active layer). An impressive PCE of >15% was reached for this blend with a 1 cm2 active layer. In general, Y7 showed improved performances over Y6, mainly due to its highest photoluminescence, lower non-radiative energy loss, and a higher Voc of 0.867 V vs 0.834 V for Y6.

Specifications

Molecular Formula C82H86Cl4N8O2S5
Linear Formula C82H86Cl4N8O2S5
Quantity 100 mg
Synonym BTP-4 Cl
Molecular Weight (g/mol) 1517.75
Percent Purity ≥99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.