Learn More
ATCC YAC-1; Lymphoma; Mouse (Mus musculus)
Catalog No. 502383207
Encompass
YAC-1 is a fibroblast cell that was isolated from the lymphoblast of a mouse with lymphoma. The cell can be used in assays with NK cells.
Catalog No. 50-238-3207 Supplier ATCC Supplier No. TIB160
YAC-1 is a fibroblast cell that was isolated from the lymphoblast of a mouse with lymphoma. The cell can be used in assays with NK cells.