Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Yadanziolide A | 95258-14-3 | 1 MG
SDP

Catalog No. 5000418672
Encompass_Preferred

Yadanziolide A is a terpenoid diterpenoid isolated from the cultivated dry seeds of Brucea javanica. It exhibits strong antiviral activities, specifically against tobacco mosaic virus with an IC50 of 5.5 μM, and also shows significant antitumor effects. It has a molecular weight of 426.41 and a molecular formula of C20H26O10.

  • Isolated from the cultivated dry seeds of Brucea javanica
  • Strong antiviral activity against tobacco mosaic virus (IC50 of 5.5 μM)
  • Exhibits significant antitumor effects
  • Molecular weight: 426.41
  • Molecular formula: C20H26O10
  • For research use only

Catalog No. 50-004-18672 Supplier Medchemexpress LLC Supplier No. HYN42101MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Yadanziolide A is a terpenoid diterpenoid isolated from the cultivated dry seeds of Brucea javanica. It exhibits strong antiviral activities, specifically against tobacco mosaic virus with an IC50 of 5.5 μM, and also shows significant antitumor effects. It has a molecular weight of 426.41 and a molecular formula of C20H26O10.

  • Isolated from the cultivated dry seeds of Brucea javanica
  • Strong antiviral activity against tobacco mosaic virus (IC50 of 5.5 μM)
  • Exhibits significant antitumor effects
  • Molecular weight: 426.41
  • Molecular formula: C20H26O10
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.