Learn More
Aviva Systems Biology Corp YAF2 Antibody - middle region (ARP31459_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31459_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against YAF2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human YAF2. Alternate names: MGC41856ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54232