Learn More
Aviva Systems Biology Corp YAF2 Antibody - middle region (ARP32676_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32676_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against YAF2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human YAF2. Alternate names: MGC41856ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54704