Learn More
Yamato Accessories for Yamato Digital Laboratory Stirrers Closed System Vacuum Adapter
Catalog No. 1450520C Shop All Yamato Scientific America Inc. Products
Click to view available options
For Use With (Equipment):
All LR-400/500 Stirrer
Catalog No. For Use With (Equipment)
14-505-20C All LR-400/500 Stirrer
1 options
Catalog No. 14-505-20C Supplier Yamato Supplier No. 231097
Allows stirring at low pressures.
For use with all Yamato Digital Laboratory Stirrers and all Fisher StedFast™ models.
  • Maintains a vacuum of 7MPa (0.05 torr) without excessive friction
  • Adapter is PTFE for excellent chemical resistance; has nitrile rubber seal
  • With standard taper 24/40 fitting for use with three-neck distilling flasks, bell jars and other vacuum glassware
Specifications
For Use With (Equipment) All LR-400/500 Stirrer
Type Vacuum Adapter