Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Yangambin | 13060-14-5 | MFCD04307767 | 5 MG
SDP

Catalog No. 5000420786
Encompass_Preferred

Yangambin is a furofuran lignan isolated from various plants. It functions as a selective PAF receptor antagonist and inhibits Ca2+ influx through voltage-gated Ca2+ channels, reducing intracellular Ca2+ in vascular smooth muscle cells, which leads to peripheral vasodilation. This compound also exhibits antiallergic and anti-inflammatory activities.

  • Selective PAF receptor antagonist
  • Inhibits Ca2+ influx through voltage-gated Ca2+ channels
  • Reduces intracellular Ca2+ in vascular smooth muscle cells
  • Demonstrates antiallergic activity (IC50 of 33.8 μM)
  • Demonstrates anti-inflammatory activity (IC50 of 37.4 μM)
  • Purity: 98.27%
  • Molecular weight: 446.49
  • Formula: C24H30O8
  • Appearance: Solid, white to off-white
  • For research use only

Catalog No. 50-004-20786 Supplier Medchemexpress LLC Supplier No. HYN42675MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Yangambin is a furofuran lignan isolated from various plants. It functions as a selective PAF receptor antagonist and inhibits Ca2+ influx through voltage-gated Ca2+ channels, reducing intracellular Ca2+ in vascular smooth muscle cells, which leads to peripheral vasodilation. This compound also exhibits antiallergic and anti-inflammatory activities.

  • Selective PAF receptor antagonist
  • Inhibits Ca2+ influx through voltage-gated Ca2+ channels
  • Reduces intracellular Ca2+ in vascular smooth muscle cells
  • Demonstrates antiallergic activity (IC50 of 33.8 μM)
  • Demonstrates anti-inflammatory activity (IC50 of 37.4 μM)
  • Purity: 98.27%
  • Molecular weight: 446.49
  • Formula: C24H30O8
  • Appearance: Solid, white to off-white
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.