Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Yangambin | 13060-14-5 | MFCD04307767 | 98.3% | 446.49 | C24H30O8 | 10 MG
SDP

Catalog No. 5000289849
Encompass_Preferred

Yangambin is a furofuran lignan supplied for laboratory research that functions as a calcium-channel inhibitor and platelet-activating factor (PAF) receptor antagonist. It reduces intracellular Ca2+ in vascular smooth muscle cells and has reported antiallergic and anti-inflammatory activity in vitro.

  • Acts as a calcium-channel inhibitor and PAF receptor antagonist.
  • Shows antiallergic and anti-inflammatory activity in vitro.
  • High purity (≈98.3%), suitable for biochemical and pharmacological assays.
  • Soluble in DMSO (50 mg/mL) and compatible with several in vivo formulations (≥1.25 mg/mL reported).
  • Furofuran lignan chemical class, C24H30O8, MW 446.49 Da.
  • Store at 4°C protected from light; in solution store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-89849 Supplier Medchemexpress LLC Supplier No. HYN426710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Yangambin is a furofuran lignan supplied for laboratory research that functions as a calcium-channel inhibitor and platelet-activating factor (PAF) receptor antagonist. It reduces intracellular Ca2+ in vascular smooth muscle cells and has reported antiallergic and anti-inflammatory activity in vitro.

  • Acts as a calcium-channel inhibitor and PAF receptor antagonist.
  • Shows antiallergic and anti-inflammatory activity in vitro.
  • High purity (≈98.3%), suitable for biochemical and pharmacological assays.
  • Soluble in DMSO (50 mg/mL) and compatible with several in vivo formulations (≥1.25 mg/mL reported).
  • Furofuran lignan chemical class, C24H30O8, MW 446.49 Da.
  • Store at 4°C protected from light; in solution store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.