Learn More
Medchemexpress LLC HY-N0919 5mg Medchemexpress, Yangonin CAS:500-62-9 Purity:>98%
SDP
Catalog No. 501970316
Encompass_Preferred
Medchemexpress, HY-N0919 5mg Yangonin CAS:500-62-9 Yangonin exhibits affinity for the human recombinant cannabinoid CB1 receptor with an IC50 and a Ki of 1.79 ± 0.53 μM and 0.72±0.21 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0316 Supplier Medchemexpress LLC Supplier No. HYN09195MG
Medchemexpress, HY-N0919 5mg Yangonin CAS:500-62-9 Yangonin exhibits affinity for the human recombinant cannabinoid CB1 receptor with an IC50 and a Ki of 1.79 ± 0.53 μM and 0.72±0.21 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.