Learn More
Med Vet International Bulb Tip Sterile Yankauers
Catalog No. 502831662
Encompass_Preferred
Bulb Tip Sterile Yankauers: Our yankauers feature slip-resistant handles for a secure grip, smooth inner surface for maximum, non-occluding flow, transparent construction for easy assessment of aspirate and shatterproof construction that helps aid patient safety. Ribbed connector for various sizes of connecting tubing. Optional control vent for suctioning control. With control vent, flexible.
Catalog No. 50-283-1662 Supplier Med Vet International Supplier No. DYND52132
Bulb Tip Sterile Yankauers: Our yankauers feature slip-resistant handles for a secure grip, smooth inner surface for maximum, non-occluding flow, transparent construction for easy assessment of aspirate and shatterproof construction that helps aid patient safety. Ribbed connector for various sizes of connecting tubing. Optional control vent for suctioning control. With control vent, flexible.