Learn More
Santa Cruz Biotechnology Anti-YAP1 Antibody (63.7), 100 ug/mL
Supplier:  Santa Cruz Biotechnology SC101199
Encompass
Recommended for detection of YAP of mouse, rat and human origin by WB, IP, IF, IHC(P) and ELISA.
Catalog No. NC9850693