Learn More
Medchemexpress LLC YAP/TAZ inhibitor-1 | 2093565-23-0 | 99.7% | 621.81 | 10 MG
SDP
Catalog No. 5000345532
Encompass_Preferred
YAP/TAZ inhibitor-1 is a YAP/TAZ inhibitor extracted from patent WO2017058716A1, Compound 1, with an IC50 of <0.100 μΜ in firefly luciferase assay. YAP and TAZ are transcriptional co-activators of the Hippo pathway network and regulate cell proliferation, migration, and apoptosis. YAP/TAZ inhibitor-1 is an inhibitor of transcriptional coactivator with PDZ binding motif/Yes-associated protein transcriptional coactivator (TAZ/YAP).
  • It is a YAP/TAZ inhibitor.
  • It is extracted from patent WO2017058716A1, compound 1.
  • It has an IC50 of <0.100 μΜ in firefly luciferase assay.
  • It inhibits transcriptional coactivator with PDZ binding motif/Yes-associated protein transcriptional coactivator (TAZ/YAP).
Catalog No. 50-003-45532 Supplier Medchemexpress LLC Supplier No. HY1114291MG
Add to Cart
Add to Cart
YAP/TAZ inhibitor-1 is a YAP/TAZ inhibitor extracted from patent WO2017058716A1, Compound 1, with an IC50 of <0.100 μΜ in firefly luciferase assay. YAP and TAZ are transcriptional co-activators of the Hippo pathway network and regulate cell proliferation, migration, and apoptosis. YAP/TAZ inhibitor-1 is an inhibitor of transcriptional coactivator with PDZ binding motif/Yes-associated protein transcriptional coactivator (TAZ/YAP).
  • It is a YAP/TAZ inhibitor.
  • It is extracted from patent WO2017058716A1, compound 1.
  • It has an IC50 of <0.100 μΜ in firefly luciferase assay.
  • It inhibits transcriptional coactivator with PDZ binding motif/Yes-associated protein transcriptional coactivator (TAZ/YAP).