Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC YAP/TAZ inhibitor-1 | 2093565-23-0 | 99.7% | 621.81 | 10 MG
SDP

Catalog No. 5000346347
Encompass_Preferred

YAP/TAZ inhibitor-1 is a YAP/TAZ inhibitor, extracted from patent WO2017058716A1, Compound 1. It has an IC50 of <0.100 μΜ in a firefly luciferase assay. As an inhibitor of transcriptional coactivator with PDZ binding motif/Yes-associated protein transcriptional coactivator (TAZ/YAP), it targets transcriptional co-activators of the Hippo pathway network, which regulate cell proliferation, migration, and apoptosis.

  • Appearance: solid
  • Color: off-white to yellow
  • Shipping: room temperature in continental US; may vary elsewhere
  • Storage (solid): 4°C, protect from light
  • Storage (in solvent): -80°C for 6 months; -20°C for 1 month (protect from light)

Catalog No. 50-003-46347 Supplier Medchemexpress LLC Supplier No. HY111429100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

YAP/TAZ inhibitor-1 is a YAP/TAZ inhibitor, extracted from patent WO2017058716A1, Compound 1. It has an IC50 of <0.100 μΜ in a firefly luciferase assay. As an inhibitor of transcriptional coactivator with PDZ binding motif/Yes-associated protein transcriptional coactivator (TAZ/YAP), it targets transcriptional co-activators of the Hippo pathway network, which regulate cell proliferation, migration, and apoptosis.

  • Appearance: solid
  • Color: off-white to yellow
  • Shipping: room temperature in continental US; may vary elsewhere
  • Storage (solid): 4°C, protect from light
  • Storage (in solvent): -80°C for 6 months; -20°C for 1 month (protect from light)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.