Learn More
Medchemexpress LLC YAP/TAZ inhibitor-2 | 2762617-31-0 | 99.8% | 390.33 | 5 MG
SDP
Catalog No. 5000362952
Encompass_Preferred
YAP/TAZ inhibitor-2 is a potent and orally active TEAD-YAP/TAZ inhibitor with an EC50 value of 3 nM. It demonstrates significant anti-proliferative and anti-tumor activities in both in vitro and in vivo studies.
  • Potent and orally active TEAD-YAP/TAZ inhibitor
  • Displays anti-proliferative activity with an IC50 of 5.33 nM in NCI-H226 cells
  • Demonstrates anti-tumor activity in BALB/c nude mice xenograft models
  • EC50 value of 3 nM
Catalog No. 50-003-62952 Supplier Medchemexpress LLC Supplier No. HY1473225MG
YAP/TAZ inhibitor-2 is a potent and orally active TEAD-YAP/TAZ inhibitor with an EC50 value of 3 nM. It demonstrates significant anti-proliferative and anti-tumor activities in both in vitro and in vivo studies.
  • Potent and orally active TEAD-YAP/TAZ inhibitor
  • Displays anti-proliferative activity with an IC50 of 5.33 nM in NCI-H226 cells
  • Demonstrates anti-tumor activity in BALB/c nude mice xenograft models
  • EC50 value of 3 nM