Learn More
Medchemexpress LLC Yap/taz inhibitor-2 | 2762617-31-0 | 99.8% | C19H14F4N4O | 10MG
SDP
Catalog No. 5000258857
Encompass_Preferred
YAP/TAZ inhibitor-2 is a small-molecule TEAD-YAP/TAZ inhibitor with potent nanomolar activity that disrupts TEAD-YAP/TAZ signaling, shows antiproliferative effects in cancer cell lines, and demonstrates oral activity in vivo. The compound is supplied as a light yellow solid with documented solubility, storage, and purity information.
  • Potent TEAD-YAP/TAZ inhibition (EC50 = 3 nM).
  • Antiproliferative activity in NCI-H226 cells (IC50 = 5.33 nM).
  • High chemical purity (99.81%).
  • Soluble in DMSO (100 mg/mL).
  • Stable as powder at -20°C for long-term storage.
  • Light yellow to yellow solid appearance.
Catalog No. 50-002-58857 Supplier Medchemexpress LLC Supplier No. HY14732210MG
YAP/TAZ inhibitor-2 is a small-molecule TEAD-YAP/TAZ inhibitor with potent nanomolar activity that disrupts TEAD-YAP/TAZ signaling, shows antiproliferative effects in cancer cell lines, and demonstrates oral activity in vivo. The compound is supplied as a light yellow solid with documented solubility, storage, and purity information.
  • Potent TEAD-YAP/TAZ inhibition (EC50 = 3 nM).
  • Antiproliferative activity in NCI-H226 cells (IC50 = 5.33 nM).
  • High chemical purity (99.81%).
  • Soluble in DMSO (100 mg/mL).
  • Stable as powder at -20°C for long-term storage.
  • Light yellow to yellow solid appearance.