Learn More
Cell Signaling Technology YAP/TAZ Transcriptional Targets Antibody Sampler Kit, Unconjugated, 1 Kit
SDP
Catalog No. 502732926
Encompass
The YAP/TAZ Transcriptional Targets Antibody Sampler Kit provides an economical solution for detecting proteins regulated by the transcriptional co-activators YAP and TAZ. These proteins act as key mediators in the Hippo signaling pathway, controlling essential cell functions such as proliferation and survival. The kit includes enough primary antibody to perform two western blot experiments per target.
  • Provides comprehensive detection of key transcriptional targets including CTGF, CYR61, IGFBP3, Integrin beta2, Survivin, Axl, Pan-TEAD, and Lamin B2.
  • Includes high-quality rabbit monoclonal antibodies validated for western blotting.
  • Supplied with an HRP-linked secondary antibody for convenient detection.
  • Optimized for studying gene expression regulated by the Hippo signaling pathway.
Catalog No. 50-273-2926 Supplier Cell Signaling Technology Supplier No. 56674T
The YAP/TAZ Transcriptional Targets Antibody Sampler Kit provides an economical solution for detecting proteins regulated by the transcriptional co-activators YAP and TAZ. These proteins act as key mediators in the Hippo signaling pathway, controlling essential cell functions such as proliferation and survival. The kit includes enough primary antibody to perform two western blot experiments per target.
  • Provides comprehensive detection of key transcriptional targets including CTGF, CYR61, IGFBP3, Integrin beta2, Survivin, Axl, Pan-TEAD, and Lamin B2.
  • Includes high-quality rabbit monoclonal antibodies validated for western blotting.
  • Supplied with an HRP-linked secondary antibody for convenient detection.
  • Optimized for studying gene expression regulated by the Hippo signaling pathway.