Learn More
Selleck Chemical LLC YAP-TEAD Inhibitor 1 (Peptide 17) 1mg
SDP
Catalog No. 501366424
Encompass_Preferred
YAP-TEAD Inhibitor 1 (Peptide 17) is a YAP-TEAD protein-protein interaction inhibitor which has potential usage in treatment of YAP-involved cancers with IC50 of 25 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6424 Supplier Selleck Chemical LLC Supplier No. S81641MG
Add to Cart
Add to Cart
YAP-TEAD Inhibitor 1 (Peptide 17) is a YAP-TEAD protein-protein interaction inhibitor which has potential usage in treatment of YAP-involved cancers with IC50 of 25 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.