Learn More
Aviva Systems Biology Corp YARS Antibody - N-terminal region (ARP40472_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40472_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against YARS. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human YARS. Alternate names: CMTDIC, TYRRS, YRS, YTSApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57472