Learn More
Electron Microscopy Sciences Yasargil Bayonet Scissors Straight
SDP
Catalog No. 5029804
Encompass_Preferred
Delicate stainless steel scissors with a spring handle; 8" (203 mm) long.
Catalog No. 50-298-04 Supplier Electron Microscopy Sciences Supplier No. 729368S
Delicate stainless steel scissors with a spring handle; 8" (203 mm) long.