Learn More
Takara Bio Yatalase™ Enzyme, 2g
SDP
Catalog No. 50444030
Encompass
Yatalase Enzyme is used to lyse the cell walls of filamentous fungi. The product is prepared from culture supernatants of Corynebacterium sp. OZ-21, and consists mainly of chitinase, chitobiase and β-1,3-glucanase. 2g
Catalog No. 50-444-030 Supplier Takara Bio Supplier No. T017
Yatalase Enzyme is used to lyse the cell walls of filamentous fungi. The product is prepared from culture supernatants of Corynebacterium sp. OZ-21, and consists mainly of chitinase, chitobiase and β-1,3-glucanase. 2g