Learn More
Medchemexpress LLC YB-0158 (Wnt pathway inhibitor 2) | 1144043-83-3 | 99.8% | 703.59 | 1 MG
SDP
Catalog No. 5000350605
Encompass_Preferred
YB-0158 is a reverse-turn peptidomimetic and a potent colorectal cancer stem cell (CSC) targeting agent. It disrupts Sam68-Src interactions and induces apoptosis in CRC cells, demonstrating anti-cancer activities.
  • Disrupts Sam68-Src interactions.
  • Induces apoptosis in CRC cells.
  • Potent colorectal cancer stem cell (CSC) targeting agent.
  • Shows anti-cancer activities.
Catalog No. 50-003-50605 Supplier Medchemexpress LLC Supplier No. HY1365411MG
YB-0158 is a reverse-turn peptidomimetic and a potent colorectal cancer stem cell (CSC) targeting agent. It disrupts Sam68-Src interactions and induces apoptosis in CRC cells, demonstrating anti-cancer activities.
  • Disrupts Sam68-Src interactions.
  • Induces apoptosis in CRC cells.
  • Potent colorectal cancer stem cell (CSC) targeting agent.
  • Shows anti-cancer activities.