Learn More
Medchemexpress LLC YB-0158 (Wnt pathway inhibitor 2) | 1144043-83-3 | 99.8% | 703.59 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13654150MG
Encompass_Preferred
YB-0158 is a reverse-turn peptidomimetic that acts as a potent colorectal cancer stem cell (CSC) targeting agent. It disrupts Sam68-Src interactions and induces apoptosis in CRC cells, demonstrating anti-cancer activities.
  • Significantly increases apoptosis in CRC cells
  • Decreases CBP recruitment at the promoter of Wnt/beta-Catenin target genes LGR5 and MYC
  • Shows an EC50 of 1.64 μM in cultured MC38 cells
  • Soluble in DMSO (100 mg/mL)
Catalog No. 50-003-49530