Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC YBX1 protein, human (His) | >90.0% | 20UG
SDP

Catalog No. 5000174037
Encompass_Preferred

Recombinant full-length human YBX1 protein with a C-terminal 6xHis tag, expressed in Escherichia coli and supplied lyophilized from PBS with 6% trehalose. Intended for research use in biochemical assays, binding studies, and detection workflows.

  • Full-length human protein with C-terminal His tag.
  • Expressed in Escherichia coli for bacterial expression systems.
  • Lyophilized formulation for long-term storage.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Predicted molecular mass 42.7 kDa; SDS-PAGE shows ~60 kDa band.
  • Stable when stored at -20°C; reconstitute and aliquot for use.

Catalog No. 50-001-74037 Supplier Medchemexpress LLC Supplier No. HYP70420520UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant full-length human YBX1 protein with a C-terminal 6xHis tag, expressed in Escherichia coli and supplied lyophilized from PBS with 6% trehalose. Intended for research use in biochemical assays, binding studies, and detection workflows.

  • Full-length human protein with C-terminal His tag.
  • Expressed in Escherichia coli for bacterial expression systems.
  • Lyophilized formulation for long-term storage.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Predicted molecular mass 42.7 kDa; SDS-PAGE shows ~60 kDa band.
  • Stable when stored at -20°C; reconstitute and aliquot for use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.