Learn More
Aviva Systems Biology Corp YBX1 Antibody - middle region (ARP34396_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34396_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against YBX1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human YBX1. Alternate names: BP-8, CSDA2, CSDB, DBPB, MDR-NF1, MGC104858, MGC110976, MGC117250, NSEP-1, NSEP1, YB-1, YB1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-55377