Learn More
Aviva Systems Biology Corp YBX2 Antibody - middle region (ARP34450_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34450_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against YBX2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human YBX2. Alternate names: DBPC, MSY2, CSDA3, CONTRINApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55398