Learn More
AdipoGen YC-1
SDP
Catalog No. 502562363
Encompass_Preferred
Chemical. CAS 170632-47-0. Formula C19H16N2O2. MW 304.4. Nitric oxide NO independent, superoxide-sensitive soluble guanylyl cyclase sGC activator. Induces concentration-dependent increase in cGMP levels. Inhibits platelet adhesion to collagen. Thombosis inhibitor. Non-specific phosphodiesterase inhibitor. Na channel blocker. Anticancer compound. HIF-alpha inhibitor. Blocks angiogenesis. Tumor growth inhibitor. Apoptosis inducer. NK cell differentiation enhancer. TRAIL sensitizer.
Catalog No. 50-256-2363 Supplier AdipoGen Supplier No. AGCR10120M001
Chemical. CAS 170632-47-0. Formula C19H16N2O2. MW 304.4. Nitric oxide NO independent, superoxide-sensitive soluble guanylyl cyclase sGC activator. Induces concentration-dependent increase in cGMP levels. Inhibits platelet adhesion to collagen. Thombosis inhibitor. Non-specific phosphodiesterase inhibitor. Na channel blocker. Anticancer compound. HIF-alpha inhibitor. Blocks angiogenesis. Tumor growth inhibitor. Apoptosis inducer. NK cell differentiation enhancer. TRAIL sensitizer.