Learn More
Abcam YC-1, guanylyl cyclase activator, 10MG
Catalog No. 502707136
Encompass_Preferred
MW 304.3 Da, Purity >99%. Reversible soluble guanylyl cyclase activator (EC₅₀ = 18.6 μM). NO-independent. Induces a concentration dependent increase in cGMP in vitro. Reduces blood-brain barrier permeability through HIF-1 inhibiton in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7136 Supplier Abcam Supplier No. AB12091510MG
MW 304.3 Da, Purity >99%. Reversible soluble guanylyl cyclase activator (EC₅₀ = 18.6 μM). NO-independent. Induces a concentration dependent increase in cGMP in vitro. Reduces blood-brain barrier permeability through HIF-1 inhibiton in vivo.
The product is subject to the following: Abcam Restricted Use Statement