Learn More
Biolog Inc YCFAC BROTH 500 mL - Yeast Casitone Fatty Acid Broth with Carbohydrates - 500 mL bottle, 1 Bottle/Pkg
SDP
Catalog No. NC1553100
Encompass
This item has a minimum order quantity of 5 bottles. Yeast Casitone Fatty Acids Broth with Carbohydrates (YCFAC BROTH) is an enriched nonselective media for the cultivation of anaerobic bacteria and other fastidious microorganisms. YCFAC-B, when handled and incubated appropriately, has been shown to recover a wide range of difficult to culture organisms from gut microbiota samples. This media is prepared, dispensed, and packaged under oxygen-free conditions to prevent the formation of oxidized products prior to use. Also available in 7ml tubes.YCFAC broth is an enriched nonselective media used in the isolation and cultivation of a wide variety of bacteria found in the human gut, including Faecalibacterium prausnitzii. The basic nutritive components of this media come from yeast extract and pancreatic digest of casein. This basal medium is then enriched with various specific vitamins, sugars, and short chain fatty acids to ensure growth of even the most fastidious gut microbes.
Catalog No. NC1553100 Supplier Biolog Inc Supplier No. AS6805
This item has a minimum order quantity of 5 bottles. Yeast Casitone Fatty Acids Broth with Carbohydrates (YCFAC BROTH) is an enriched nonselective media for the cultivation of anaerobic bacteria and other fastidious microorganisms. YCFAC-B, when handled and incubated appropriately, has been shown to recover a wide range of difficult to culture organisms from gut microbiota samples. This media is prepared, dispensed, and packaged under oxygen-free conditions to prevent the formation of oxidized products prior to use. Also available in 7ml tubes.YCFAC broth is an enriched nonselective media used in the isolation and cultivation of a wide variety of bacteria found in the human gut, including Faecalibacterium prausnitzii. The basic nutritive components of this media come from yeast extract and pancreatic digest of casein. This basal medium is then enriched with various specific vitamins, sugars, and short chain fatty acids to ensure growth of even the most fastidious gut microbes.