Learn More
Biolog Inc YCFAC-B - Yeast Castione Fatty Acid Agar with Carbohydrates and Blood, 4 Plates/Pkg
SDP
Catalog No. NC1420048
Encompass
This item has a minimum qty of 25 per supplier requirements.
Yeast Casitone Fatty Acids Agar with Carbohydrates & Sheep Blood (YCFAC-B) is an enriched nonselective media for the cultivation of anaerobic bacteria and other fastidious microorganisms. YCFAC-B, when handled and incubated appropriately, has been shown to recover a wide range of difficult to culture organisms from gut microbiota samples. This media is prepared, dispensed, and packaged under oxygen-free conditions to prevent the formation of oxidized products prior to use.YCFAC-B agar is an enriched nonselective media used in the isolation and cultivation of a wide variety of bacteria found in the human gut, including Faecalibacterium prausnitzii. The basic nutritive components of this media come from yeast extract and pancreatic digest of casein.
This item is not returnable. View return policy
Catalog No. NC1420048 Supplier Biolog Inc Supplier No. AS677
This item has a minimum qty of 25 per supplier requirements.
Yeast Casitone Fatty Acids Agar with Carbohydrates & Sheep Blood (YCFAC-B) is an enriched nonselective media for the cultivation of anaerobic bacteria and other fastidious microorganisms. YCFAC-B, when handled and incubated appropriately, has been shown to recover a wide range of difficult to culture organisms from gut microbiota samples. This media is prepared, dispensed, and packaged under oxygen-free conditions to prevent the formation of oxidized products prior to use.YCFAC-B agar is an enriched nonselective media used in the isolation and cultivation of a wide variety of bacteria found in the human gut, including Faecalibacterium prausnitzii. The basic nutritive components of this media come from yeast extract and pancreatic digest of casein.