Learn More
Biolog Inc YCFAC BROTH - Yeast Casitone Fatty Acid Broth with Carbohydrates, 10 Tubes/Pkg
SDP
Catalog No. NC1383459
Encompass
This item has a minimum qty of 2 per supplier requirements.
Yeast Casitone Fatty Acids Broth with Carbohydrates (YCFAC BROTH) is an enriched nonselective media for the cultivation of anaerobic bacteria & other fastidious microorganisms. YCFAC-B, when handled & incubated appropriately, has been shown to recover a wide range of difficult to culture organisms from gut microbiota samples. This media is prepared, dispensed, & packaged under oxygen-free conditions to prevent the formation of oxidized products prior to use. Also available in 500ml bottles.YCFAC broth is an enriched nonselective media used in the isolation & cultivation of a wide variety of bacteria found in the human gut, including Faecalibacterium prausnitzii. The basic nutritive components of this media come from yeast extract & pancreatic digest of casein.
Catalog No. NC1383459 Supplier Biolog Inc Supplier No. AS680
This item has a minimum qty of 2 per supplier requirements.
Yeast Casitone Fatty Acids Broth with Carbohydrates (YCFAC BROTH) is an enriched nonselective media for the cultivation of anaerobic bacteria & other fastidious microorganisms. YCFAC-B, when handled & incubated appropriately, has been shown to recover a wide range of difficult to culture organisms from gut microbiota samples. This media is prepared, dispensed, & packaged under oxygen-free conditions to prevent the formation of oxidized products prior to use. Also available in 500ml bottles.YCFAC broth is an enriched nonselective media used in the isolation & cultivation of a wide variety of bacteria found in the human gut, including Faecalibacterium prausnitzii. The basic nutritive components of this media come from yeast extract & pancreatic digest of casein.