Learn More
Zymo Research Corporation YD Lysis Buffer, 4.8ml
SDP
Catalog No. NC9527625
Encompass_Preferred
YD Lysis Buffer provides a reliable and efficient method for the isolation of Genomic DNA from a broad spectrum of fungus species using Zymolyase and Zymo-Spin column purification technology.
Catalog No. NC9527625 Supplier Zymo Research Corporation Supplier No. D20022
YD Lysis Buffer provides a reliable and efficient method for the isolation of Genomic DNA from a broad spectrum of fungus species using Zymolyase and Zymo-Spin column purification technology.