Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Yeast, active, dry
SCI_ED

Catalog No. s25632 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Form:
Powder
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
S25632 100 g Powder
S25632A 500 g Powder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25632
Only null left
Add to Cart
Add to Cart

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 8013-01-2

Specifications

Color Tan
Form Powder
Quantity 100 g
Chemical Name or Material Yeast, Active, Dry
ShelfLife 2 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.