Learn More
Bioworld Yeast Agar, 500 g
Catalog No. 50254480
Encompass_Preferred
Yeast AgarUsed for the selective isolation of dermatophytes and other pathogenic fungi from routine clinical specimenspH: 6.6Dehydrated Appearance: homogeneous powderPrepared Appearance: yellow to tan agarSoluble in boiling waterContains:Glucose SoyPeptone Malt Extract Yeast Extract Agar
Catalog No. 50-254-480 Supplier Bioworld Supplier No. 306260081
Yeast AgarUsed for the selective isolation of dermatophytes and other pathogenic fungi from routine clinical specimenspH: 6.6Dehydrated Appearance: homogeneous powderPrepared Appearance: yellow to tan agarSoluble in boiling waterContains:Glucose SoyPeptone Malt Extract Yeast Extract Agar