Learn More
Bioworld Yeast Extract Glucose Broth, 500 g
Catalog No. 50254481
Encompass_Preferred
Yeast Extract Glucose BrothMedium for the cultivation and maintanace of Bacillus licheniformis, Bacillus species, Clavibacter michiganense, Flavobacterium indologenes, Hafnia alvei, Pseudomonas fluorescens, and Serratia marcescens, as well as yeast spp.Soluble in waterContains:Glucose - 15g/LPotassium Phosphate Dibasic - 5.2g/LPotassium Phosphate Monobasic - 3.18g/L Magnesium Sulphate - 0.12g/LYeast Extract - 0.5g/LAmmonium Chloride - 0.54g/L
Catalog No. 50-254-481 Supplier Bioworld Supplier No. 306260091
Yeast Extract Glucose BrothMedium for the cultivation and maintanace of Bacillus licheniformis, Bacillus species, Clavibacter michiganense, Flavobacterium indologenes, Hafnia alvei, Pseudomonas fluorescens, and Serratia marcescens, as well as yeast spp.Soluble in waterContains:Glucose - 15g/LPotassium Phosphate Dibasic - 5.2g/LPotassium Phosphate Monobasic - 3.18g/L Magnesium Sulphate - 0.12g/LYeast Extract - 0.5g/LAmmonium Chloride - 0.54g/L