Learn More
HIMEDIA Yeast Carbon Base, M141, Microbiology Culture Media, Dehydrated Culture Media, Specialty Medium, Yeast Identification, Nitrogen Assimilation, Powder, 100 g
Catalog No. NC4013688
Encompass_Preferred
Yeast Carbon Base is the defined medium for classifying yeasts by their ability to assimilate individual nitrogen compounds. It supplies dextrose as the carbon source together with a complete background of amino acids, vitamins, trace elements and mineral salts, so the analyst varies only the nitrogen source under test. This makes it a standard reagent in yeast taxonomy and identification, complementing Yeast Nitrogen Base in auxanographic assimilation studies. Dehydrated powder, store in a cool dry place. Packaged as 100 g.
Catalog No. NC4013688 Supplier HIMEDIA Supplier No. M141100G
Yeast Carbon Base is the defined medium for classifying yeasts by their ability to assimilate individual nitrogen compounds. It supplies dextrose as the carbon source together with a complete background of amino acids, vitamins, trace elements and mineral salts, so the analyst varies only the nitrogen source under test. This makes it a standard reagent in yeast taxonomy and identification, complementing Yeast Nitrogen Base in auxanographic assimilation studies. Dehydrated powder, store in a cool dry place. Packaged as 100 g.