Learn More
MABOGEN LLC Yeast Display Vectors, high copy (display or secretion)
Catalog No. NC4001093
Encompass
YSD-C-2u and pYSD-N-2u are yeast display vectors for scFv display on the surface of yeast strain EBY100. pYSD-C/N-2u displays scFv fused to the C/N terminal of AGA2 through GS linker
Catalog No. NC4001093 Supplier MABOGEN LLC Supplier No. YD002
YSD-C-2u and pYSD-N-2u are yeast display vectors for scFv display on the surface of yeast strain EBY100. pYSD-C/N-2u displays scFv fused to the C/N terminal of AGA2 through GS linker