Learn More
NEOGEN Yeast Extract Dextrose Chloramphenicol Agar, 5 kg
SDP
Catalog No. 502015682
Encompass_Preferred
Yeast Extract Dextrose Chloramphenicol Agar is a selective medium for the enumeration of yeasts and molds in milk and other dairy products, and is not intended for use in the diagnosis of disease or other conditions in humans. The medium is said to have superior storage properties to O.G.Y.E. and also has the advantage of incorporating an autoclavable supplement.For more information visit https://www.neogen.com/solutions/Microbiology/yeast-extract-dextrose-chloramphenicol-agar
Catalog No. 50-201-5682 Supplier NEOGEN Supplier No. 700004590
Yeast Extract Dextrose Chloramphenicol Agar is a selective medium for the enumeration of yeasts and molds in milk and other dairy products, and is not intended for use in the diagnosis of disease or other conditions in humans. The medium is said to have superior storage properties to O.G.Y.E. and also has the advantage of incorporating an autoclavable supplement.For more information visit https://www.neogen.com/solutions/Microbiology/yeast-extract-dextrose-chloramphenicol-agar