Learn More
NEOGEN Yeast Extract, 10 kg
SDP
Catalog No. 502015764
Encompass_Preferred
Yeast Extract is an autolysate of yeast cells used in preparing microbiological culture media in a laboratory setting. Yeast Extract is not intended for use in the diagnosis of disease or other conditions in humans.Yeast Extract is the water-soluble portion of autolyzed yeast. The autolysis is carefully controlled to preserve naturally occurring B-complex vitamins. Yeast Extract is prepared and standardized for bacteriological use and cell cultures and is an excellent stimulator of bacterial growth. Yeast Extract is generally employed in the concentration of 0.3% - 0.5%. Yeast Extract is typically prepared by growing baker’s yeast, Saccharomyces spp., in a carbohydrate-rich plant medium. The yeast is harvested, washed, and resuspended in water, where it undergoes autolysis, i.e., self-digestion using the yeast’s enzymes.For more information visit https://www.neogen.com/solutions/Microbiology/yeast-extract
Catalog No. 50-201-5764 Supplier NEOGEN Supplier No. 700003546
Yeast Extract is an autolysate of yeast cells used in preparing microbiological culture media in a laboratory setting. Yeast Extract is not intended for use in the diagnosis of disease or other conditions in humans.Yeast Extract is the water-soluble portion of autolyzed yeast. The autolysis is carefully controlled to preserve naturally occurring B-complex vitamins. Yeast Extract is prepared and standardized for bacteriological use and cell cultures and is an excellent stimulator of bacterial growth. Yeast Extract is generally employed in the concentration of 0.3% - 0.5%. Yeast Extract is typically prepared by growing baker’s yeast, Saccharomyces spp., in a carbohydrate-rich plant medium. The yeast is harvested, washed, and resuspended in water, where it undergoes autolysis, i.e., self-digestion using the yeast’s enzymes.For more information visit https://www.neogen.com/solutions/Microbiology/yeast-extract