Learn More
Sigma Aldrich Fine Chemicals Biosciences Yeast Extract suitable for microbiology | 8013-01-2 | MFCD00132599 | 500G
Catalog No. NC0720493
Encompass
Yeast Extract suitable for microbiology | Purity: >=10% total nitrogen (N) basis; >=4.5% amino-nitrogen basis | 8013-01-2 | MFCD00132599 | 500G
Catalog No. NC0720493 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 92144500GF
Yeast Extract suitable for microbiology | Purity: >=10% total nitrogen (N) basis; >=4.5% amino-nitrogen basis | 8013-01-2 | MFCD00132599 | 500G