Learn More
Sigma Aldrich Fine Chemicals Biosciences Yeast Extract, 8013-01-2, MFCD00132599, 500 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 70161500G
Encompass
For microbiology. A water soluble extract of autolyzed yeast cells. Yeast extract is a mixture of amino acids, peptides, water soluble vitamins and carbohydrates and can be used as additive for culture media.
Catalog No. NC0629501