Learn More
Bioworld Yeast Extract, 100 g
Supplier:  Bioworld 306200961
Encompass_Preferred
Yeast ExtractAqueous extract of autolysed brewer's yeast Has a high vitamin content, providing excellent growth factors for most microorganismsIs usually added to culture media to provide a source of vitamins at a concentration of 3-5 g/LAfter opening, tightly recap and place the container in a low humidity environment at room temperature, also protect from strong light sources. The dehydrated powder should be discarded if it is not free flowing and the color has changed from the original light beige. Specifications:Total Solids: 96.0 ± 2.0% Amino Nitrogen/Total Nitrogen %: 28-46%Protein %: 62-74%Ash: ≤14%pH (5% Solution, 25°C): 6.2-7.0Total Plate Count (CFU/g): <10000 CFU/gYeast and Mold: <100Salmonella : Negative Coliforms: NegativeE.Coli: Negative
Catalog No. 50-254-394