Learn More
LAMDA BIOTECH INC Yeast Extract
Catalog No. 502045831
Encompass_Preferred
High quality Yeast Extract. This Yeast Extract contains a mixture of vitamins, amino acids, peptides and carbohydrates. It is widely used for general bacteriological research work with a variety of microorganisms, such as for making various microbiological media and as an additive for other culture media.
Catalog No. 50-204-5831 Supplier LAMDA BIOTECH INC Supplier No. C108
High quality Yeast Extract. This Yeast Extract contains a mixture of vitamins, amino acids, peptides and carbohydrates. It is widely used for general bacteriological research work with a variety of microorganisms, such as for making various microbiological media and as an additive for other culture media.