Learn More
NEOGEN Yeast Extract Agar, 5 kg
SDP
Catalog No. 502015355
Encompass_Preferred
Yeast Extract Agar is used for the enumeration of microorganisms in potable and freshwater samples and is not intended for use in the diagnosis of disease or other conditions in humans. A nutrient agar corresponding to the standard formulation for the plate count of micro-organisms in water and dairy products. This medium is also useful for teaching and demonstration purposes using non-fastidious organisms.For more information visit https://www.neogen.com/solutions/Microbiology/yeast-extract-agar
Catalog No. 50-201-5355 Supplier NEOGEN Supplier No. 700004449
Yeast Extract Agar is used for the enumeration of microorganisms in potable and freshwater samples and is not intended for use in the diagnosis of disease or other conditions in humans. A nutrient agar corresponding to the standard formulation for the plate count of micro-organisms in water and dairy products. This medium is also useful for teaching and demonstration purposes using non-fastidious organisms.For more information visit https://www.neogen.com/solutions/Microbiology/yeast-extract-agar