Learn More
Research Products International Corp Yeast Extract, 1 Gram Capsules, 1 Kilogram
SDP
Catalog No. 50489154
Encompass_Preferred
Each capsule contains 1g Yeast Extract.
Convenient and easy to use - simply add required number of premeasured capsules according to formulation being prepared without weighing or measuring. Water must be 37°C or warmer to completely dissolve gelatin capsule.
Catalog No. 50-489-154 Supplier Research Products International Corp Supplier No. Y200261000.0
Each capsule contains 1g Yeast Extract.
Convenient and easy to use - simply add required number of premeasured capsules according to formulation being prepared without weighing or measuring. Water must be 37°C or warmer to completely dissolve gelatin capsule.