Learn More
Research Products International Corp Yeast Extract, Granulated, 10 KG
SDP
Catalog No. 502230470
Encompass_Preferred
Dried yeast autolysate with a high content of amino nitrogen and water soluble B complex vitamins. Material is specially granulated to reduce dust and increase solubility rate.
Suitable for Microbiology Applications.
Catalog No. 50-223-0470 Supplier Research Products International Corp Supplier No. Y2002510000.0
Dried yeast autolysate with a high content of amino nitrogen and water soluble B complex vitamins. Material is specially granulated to reduce dust and increase solubility rate.
Suitable for Microbiology Applications.